Di-tert-butyl hydrazodicarboxylate 98%

Lieferant: Apollo Scientific
Achtung

OR917632-100G OR917632-25G
APOSOR917632-100GEA 71.2 EUR
APOSOR917632-100G APOSOR917632-25G
Di-tert-butyl hydrazodicarboxylate 98%
Di-tert-butyl hydrazodicarboxylate
Formel: C₁₀H₂₀N₂O₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00015000
CAS-Nummer: 16466-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR