N-tert-Butylethylamine 95%
Lieferant: Apollo Scientific
OR916414-1G
OR916414-5G
APOSOR916414-1GEA
55.8
EUR
APOSOR916414-1G
APOSOR916414-5G
N-tert-Butylethylamine 95%
N-tert-Butylethylamine
Formel:
C₆H₁₅N Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00059367 CAS-Nummer: 4432-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...