4'-(1-Pyrrolidinyl)acetophenon 96%

Lieferant: Apollo Scientific

OR914528-100G OR914528-25G OR914528-5G
APOSOR914528-100GEA 574 EUR
APOSOR914528-100G APOSOR914528-25G APOSOR914528-5G
4'-(1-Pyrrolidinyl)acetophenon 96%
4'-(1-Pyrrolidinyl)acetophenon
Formel: C₁₂H₁₅NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01075691
CAS-Nummer: 21557-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR