(2-Imidazol-1-yl-phenyl)methanol 95%

Lieferant: Apollo Scientific

OR913124-10G OR913124-1G OR913124-5G
APOSOR913124-10GEA 906 EUR
APOSOR913124-10G APOSOR913124-1G APOSOR913124-5G
(2-Imidazol-1-yl-phenyl)methanol 95%
(2-Imidazol-1-yl-phenyl)methanol
Formel: C₁₀H₁₀N₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD05864823
CAS-Nummer: 25373-56-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR