6-Nitro-1,2,3,4-tetrahydroquinoxaline 95%
Lieferant: Apollo Scientific
OR912904-10G
OR912904-1G
OR912904-5G
APOSOR912904-10GEA
346
EUR
APOSOR912904-10G
APOSOR912904-1G
APOSOR912904-5G
6-Nitro-1,2,3,4-tetrahydroquinoxaline 95%
6-Nitro-1,2,3,4-tetrahydroquinoxaline
Formel:
C₈H₉N₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03788188 CAS-Nummer: 41959-35-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...