2-Hydroxycyclohexancarbonsäure 95%
Lieferant: Apollo Scientific
OR912806-1G
OR912806-250MG
APOSOR912806-1GEA
632
EUR
APOSOR912806-1G
APOSOR912806-250MG
2-Hydroxycyclohexancarbonsäure 95%
2-Hydroxycyclohexanecarboxylic acid
Formel:
C₇H₁₂O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02684437 CAS-Nummer: 609-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...