1-Methyl-1H-benzimidazole-2-thiol 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl-2-mercaptobenzimidazole
OR912702-1G
OR912702-5G
APOSOR912702-1GEA
65.4
EUR
APOSOR912702-1G
APOSOR912702-5G
1-Methyl-1H-benzimidazole-2-thiol 95%
1-Methyl-1H-benzimidazole-2-thiol
Formel:
C₈H₈N₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01788333 CAS-Nummer: 2360-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...