1-Fmoc-4-piperidineacetic acid 97%
Lieferant: Apollo Scientific
Synonyme:
2-(1-(((9H-Fluoren-9-yl)methoxy)carbonyl)piperidin-4-yl)acetic acid, Fmoc-4-piperidylacetic acid
OR912176-1G
OR912176-5G
APOSOR912176-1GEA
69.6
EUR
APOSOR912176-1G
APOSOR912176-5G
1-Fmoc-4-piperidineacetic acid 97%
1-Fmoc-4-piperidineacetic acid
Formel:
C₂₂H₂₃NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00273478 CAS-Nummer: 180181-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...