4-(Methylthio)phenylessigsäure 95%
Lieferant: Apollo Scientific
OR911637-1G
OR911637-5G
APOSOR911637-1GEA
21.5
EUR
APOSOR911637-1G
APOSOR911637-5G
4-(Methylthio)phenylessigsäure 95%
4-(Methylthio)phenylessigsäure
Formel:
C₉H₁₀O₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00192325 CAS-Nummer: 16188-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...