Fenofibric acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-(4-Chlorobenzoyl)phenoxy)-2-methylpropanoic acid
OR911610-100G
OR911610-25G
OR911610-5G
APOSOR911610-100GEA
582
EUR
APOSOR911610-100G
APOSOR911610-25G
APOSOR911610-5G
Fenofibric acid 98%
Fenofibric acid
Formel:
C₁₇H₁₅ClO₄ Schmelzpunkt: 187 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00792461 CAS-Nummer: 42017-89-0 UN-Nummer: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...