D-Tyrosine methyl ester hydrochloride 98+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
H-D-Tyr-OMe.HCl, H-D-Tyr-OMe·HCl
OR911434-100G
OR911434-500G
OR911434-25G
APOSOR911434-100GEA
111
EUR
APOSOR911434-100G
APOSOR911434-500G
APOSOR911434-25G
D-Tyrosine methyl ester hydrochloride 98+%
D-Tyrosine methyl ester hydrochloride
Formel:
C₁₀H₁₄ClNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066124 CAS-Nummer: 3728-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...