2-Nitro-p-cymene 95%

Lieferant: Apollo Scientific

OR911432-25G OR911432-5G
APOSOR911432-25GEA 85.9 EUR
APOSOR911432-25G APOSOR911432-5G
2-Nitro-p-cymene 95%
2-Nitro-p-cymene
Formel: C₁₀H₁₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007176
CAS-Nummer: 943-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR