4-Amino-N,N-diethylbenzamide 98%

Lieferant: Apollo Scientific

Synonyme: N,N-Diethyl 4-aminobenzamide

OR911374-1G OR911374-5G
APOSOR911374-1GEA 88.6 EUR
APOSOR911374-1G APOSOR911374-5G
4-Amino-N,N-diethylbenzamide 98%
4-Amino-N,N-diethylbenzamide
Formel: C₁₁H₁₆N₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00461426
CAS-Nummer: 51207-85-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR