4-Ethoxybenzamid 95%

Lieferant: Apollo Scientific

OR911170-25G OR911170-5G
APOSOR911170-25GEA 270 EUR
APOSOR911170-25G APOSOR911170-5G
4-Ethoxybenzamid 95%
4-Ethoxybenzamid
Formel: C₉H₁₁NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007996
CAS-Nummer: 55836-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR