Mth-Glycine 96%

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-2-thioxoimidazolidin-4-one

OR910252-1G OR910252-5G
APOSOR910252-1GEA 174 EUR
APOSOR910252-1G APOSOR910252-5G
Mth-Glycine 96%
3-Methyl-2-thiohydantoin
Formel: C₄H₆N₂OS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00038425
CAS-Nummer: 694-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR