6-Hydroxycumarin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Hydroxy-2-benzopyrone, 6-Hydroxy-2H-chromen-2-one
OR9098-25G
OR9098-5G
OR9098-250MG
OR9098-1G
OR9098-10G
APOSOR9098-25GEA
654
EUR
APOSOR9098-25G
APOSOR9098-5G
APOSOR9098-250MG
APOSOR9098-1G
APOSOR9098-10G
6-Hydroxycumarin 97%
6-Hydroxycumarin
Formel:
C₉H₆O₃ Schmelzpunkt: 249…253 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00630040 CAS-Nummer: 6093-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...