1-Propyl-1H-pyrazole-4-carbaldehyde 95%

Lieferant: Apollo Scientific

OR909354-1G OR909354-250MG
APOSOR909354-1GEA 111 EUR
APOSOR909354-1G APOSOR909354-250MG
1-Propyl-1H-pyrazole-4-carbaldehyde 95%
1-Propyl-1H-pyrazole-4-carbaldehyde
Formel: C₇H₁₀N₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03727244
CAS-Nummer: 473249-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR