6-Chlor-2,5-dimethylbenzimidazol 97%

Lieferant: Apollo Scientific

OR909350-1G OR909350-25G OR909350-5G
APOSOR909350-1GEA 109 EUR
APOSOR909350-1G APOSOR909350-25G APOSOR909350-5G
6-Chlor-2,5-dimethylbenzimidazol 97%
6-Chlor-2,5-dimethylbenzimidazol
Formel: C₉H₉ClN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD15143570
CAS-Nummer: 221548-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR