Ethyl-2-methoxyisonicotinat 96%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-methoxyisonicotinate
OR907546-1G
OR907546-5G
APOSOR907546-1GEA
53.8
EUR
APOSOR907546-1G
APOSOR907546-5G
Ethyl-2-methoxyisonicotinat 96%
Ethyl-2-methoxyisonicotinat
Formel:
C₉H₁₁NO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11504865 CAS-Nummer: 105596-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...