β-Methyl-γ-octanolactone (mixture of isomers) 97%

Lieferant: Apollo Scientific

Synonyme: 5-Butyl-4-methyldihydro-2(3H)-furanone

OR906422-100G OR906422-25G
APOSOR906422-100GEA 406 EUR
APOSOR906422-100G APOSOR906422-25G
β-Methyl-γ-octanolactone (mixture of isomers) 97%
beta-Methyl-gamma-octanolactone (mixture of isomers)
Formel: C₉H₁₆O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00209540
CAS-Nummer: 39212-23-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR