4-(Methoxymethyl)-5-methylisoxazole-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR906145-1G OR906145-250MG
APOSOR906145-1GEA 864 EUR
APOSOR906145-1G APOSOR906145-250MG
4-(Methoxymethyl)-5-methylisoxazole-3-carboxylic acid 95%
4-(Methoxymethyl)-5-methylisoxazole-3-carboxylic acid
Formel: C₇H₉NO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06805582
CAS-Nummer: 934156-84-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR