(S)-tert-Butyl-3-hydroxybutanoate 95%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (S)-(+)-3-hydroxybutyrate

OR905870-1G OR905870-5G
APOSOR905870-1GEA 232 EUR
APOSOR905870-1G APOSOR905870-5G
(S)-tert-Butyl-3-hydroxybutanoate 95%
(S)-tert-Butyl-3-hydroxybutanoate
Formel: C₈H₁₆O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00040559
CAS-Nummer: 82578-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR