5-Chloro-2-methoxyisonicotinic acid 98%
Lieferant: Apollo Scientific
OR904765-1G
OR904765-250MG
APOSOR904765-1GEA
545
EUR
APOSOR904765-1G
APOSOR904765-250MG
5-Chloro-2-methoxyisonicotinic acid 98%
5-Chloro-2-methoxyisonicotinic acid
Formel:
C7H6ClNO3 Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
88912-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...