Cyclopropyl-phenylketon 95%

Lieferant: Apollo Scientific

OR903882-100G OR903882-25G
APOSOR903882-100GEA 323 EUR
APOSOR903882-100G APOSOR903882-25G
Cyclopropyl-phenylketon 95%
Cyclopropyl phenyl ketone
Formel: C₁₀H₁₀O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00001294
CAS-Nummer: 3481-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR