2-Benzofuranacetic acid 95%

Lieferant: Apollo Scientific

OR903609-1G OR903609-250MG
APOSOR903609-1GEA 187 EUR
APOSOR903609-1G APOSOR903609-250MG
2-Benzofuranacetic acid 95%
2-Benzofuranacetic acid
Formel: C₁₀H₈O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00052742
CAS-Nummer: 62119-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR