L(+)-Methyl-tert-leucinat Hydrochlorid 97%
Lieferant: Apollo Scientific
Synonyme:
L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl
OR903118-100G
OR903118-25G
OR903118-5G
APOSOR903118-100GEA
405
EUR
APOSOR903118-100G
APOSOR903118-25G
APOSOR903118-5G
L(+)-Methyl-tert-leucinat Hydrochlorid 97%
L(+)-Methyl-tert-leucinat Hydrochlorid
Formel:
C₇H₁₅NO₂·HCl Molekulargewicht: 181.7 g/mol Schmelzpunkt: 183 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00040604 CAS-Nummer: 63038-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...