2,5-Dichloro-3-methoxypyridine 98%

Lieferant: Apollo Scientific

OR901752-1G OR901752-250MG
APOSOR901752-1GEA 574 EUR
APOSOR901752-1G APOSOR901752-250MG
2,5-Dichloro-3-methoxypyridine 98%
2,5-Dichloro-3-methoxypyridine
Formel: C₆H₅Cl₂NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13185540
CAS-Nummer: 1214366-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR