3',4'-Dimethoxy-2'-hydroxyacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
OR8813-25G
OR8813-5G
OR8813-1G
APOSOR8813-25GEA
163
EUR
APOSOR8813-25G
APOSOR8813-5G
APOSOR8813-1G
3',4'-Dimethoxy-2'-hydroxyacetophenon
3',4'-Dimethoxy-2'-hydroxyacetophenon
Formel:
C₁₀H₁₂O₄ Schmelzpunkt: 75…77 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00016684 CAS-Nummer: 5396-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...