3-(4-Nitrophenyl)propansäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Nitrohydrocinnamic Acid, 3-(4-Nitrophenyl)propanoic acid
OR8590-1G
OR8590-25G
OR8590-5G
APOSOR8590-1GEA
14.7
EUR
APOSOR8590-1G
APOSOR8590-25G
APOSOR8590-5G
3-(4-Nitrophenyl)propansäure
3-(4-Nitrophenyl)propionic acid
Formel:
C₉H₉NO₄ Schmelzpunkt: 164…168 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00126834 CAS-Nummer: 16642-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...