3-Brom-2-(4-morpholino)pyridin

Lieferant: Apollo Scientific

OR8533-1G OR8533-5G
APOSOR8533-1GEA 52.4 EUR
APOSOR8533-1G APOSOR8533-5G
3-Brom-2-(4-morpholino)pyridin
3-Brom-2-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00498942
CAS-Nummer: 54231-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR