3-(Dimethylamino)-1-(4-nitrophenyl)-2-propen-1-one

Lieferant: Apollo Scientific

OR8506-1G OR8506-5G
APOSOR8506-1GEA 46.4 EUR
APOSOR8506-1G APOSOR8506-5G
3-(Dimethylamino)-1-(4-nitrophenyl)-2-propen-1-one
3-(Dimethylamino)-1-(4-nitrophenyl)-2-propen-1-one
Formel: C₁₁H₁₂N₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00121191
CAS-Nummer: 68760-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR