5-Nitroindolin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-5-nitro-(1H)-indole
OR8333-10G
OR8333-1G
APOSOR8333-10GEA
42.3
EUR
APOSOR8333-10G
APOSOR8333-1G
5-Nitroindolin
5-Nitroindolin
Formel:
C₈H₈N₂O₂ Schmelzpunkt: 92…94 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005709 CAS-Nummer: 32692-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...