3-Brom-N-ethyl-5-nitrobenzamid

Lieferant: Apollo Scientific

OR8312-1G OR8312-5G
APOSOR8312-1GEA 132 EUR
APOSOR8312-1G APOSOR8312-5G
3-Brom-N-ethyl-5-nitrobenzamid
3-Brom-N-ethyl-5-nitrobenzamid
Formel: C₉H₉BrN₂O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09258762
CAS-Nummer: 929000-30-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR