4-(1H-1,2,4-Triazol-1-yl)anilin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1,2,4-Triazol-1-yl)aniline, 1-(4-Aminophenyl)-1,2,4-triazole

OR8182-100G OR8182-25G OR8182-5G
APOSOR8182-100GEA 691 EUR
APOSOR8182-100G APOSOR8182-25G APOSOR8182-5G
4-(1H-1,2,4-Triazol-1-yl)anilin 95%
4-(1H-1,2,4-Triazol-1-yl)anilin
Formel: C₈H₈N₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00171470
CAS-Nummer: 6523-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR