1-(Fmoc-amino)cyclopropanecarboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Aminocyclopropane-1-carboxylic acid, N-FMOC protected, Fmoc-ACPC-OH, Fmoc-1-aminocyclopropane-1-carboxylic acid, 1-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)cyclopropanecarboxylic acid
OR8098-1G
OR8098-250MG
APOSOR8098-1GEA
65.4
EUR
APOSOR8098-1G
APOSOR8098-250MG
1-(Fmoc-amino)cyclopropanecarboxylic acid 97%
1-(Fmoc-amino)cyclopropanecarboxylic acid
Formel:
C₁₉H₁₇NO₄ Schmelzpunkt: 225…226 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00190873 CAS-Nummer: 126705-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...