4-(1H-Benzimidazol-1-ylmethyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

OR8017-1G OR8017-5G
APOSOR8017-1GEA 109 EUR
APOSOR8017-1G APOSOR8017-5G
4-(1H-Benzimidazol-1-ylmethyl)benzoic acid 97%
4-(1H-Benzimidazol-1-ylmethyl)benzoic acid
Formel: C₁₅H₁₂N₂O₂
Schmelzpunkt: 260…265 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02180372
CAS-Nummer: 139742-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR