1-Iodo-3,5-bis(methylsulphonyl)benzene 95+%
Lieferant: Apollo Scientific
OR7988-1G
OR7988-5G
OR7988-25G
APOSOR7988-1GEA
166
EUR
APOSOR7988-1G
APOSOR7988-5G
APOSOR7988-25G
1-Iodo-3,5-bis(methylsulphonyl)benzene 95+%
1-Iodo-3,5-bis(methylsulphonyl)benzene
Formel:
C₈H₉IO₄S₂ Schmelzpunkt: 212 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04037973 CAS-Nummer: 849035-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...