6-Amino-3-brom-2-methylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Amino-3-bromo-2-picoline, 6-AMINO-3-BROMO-2-METHYLPYRIDINE, 3-Bromo-2-methyl-6-pyridylamine, 5-Bromo-6-methylpyridin-2-amine, 3-Bromo-2-methylpyridin-6-amine, 2-Amino-5-bromo-6-picoline, ['6-Amino-3-bromo-2-picoline'], 5-Bromo-6-methyl-2-pyridylamine, 2-Amino-5-bromo-6-methylpyridine
OR7864-100G
OR7864-25G
OR7864-500G
APOSOR7864-100GEA
190
EUR
APOSOR7864-100G
APOSOR7864-25G
APOSOR7864-500G
6-Amino-3-brom-2-methylpyridin
6-Amino-3-brom-2-methylpyridin
Formel:
C₆H₇BrN₂ Schmelzpunkt: 74…79 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00068230 CAS-Nummer: 42753-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...