3,5-Bis(methylsulphonyl)benzoic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR7822-1G OR7822-5G OR7822-10G
APOSOR7822-1GEA 235 EUR
APOSOR7822-1G APOSOR7822-5G APOSOR7822-10G
3,5-Bis(methylsulphonyl)benzoic acid 95+%
3,5-Bis(methylsulphonyl)benzoic acid
Formel: C₉H₁₀O₆S₂
Schmelzpunkt: 283…285 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04037979
CAS-Nummer: 90536-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR