2,3-Dimethyl-6-nitrochinoxalin
Lieferant: Apollo Scientific
|
Achtung
|
OR7790-1G
OR7790-2.5G
APOSOR7790-1GEA
46.4
EUR
APOSOR7790-1G
APOSOR7790-2.5G
2,3-Dimethyl-6-nitrochinoxalin
2,3-Dimethyl-6-nitroquinoxaline
Formel:
C₁₀H₉N₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00205436 CAS-Nummer: 2942-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...