Ethylpyruvat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester

OR7781-100G OR7781-500G
APOSOR7781-100GEA 65.5 EUR
APOSOR7781-100G APOSOR7781-500G
Ethylpyruvat
Ethylpyruvat

Material for Pharmaceuticals and agrochemicals; Electronics solvent; Intermediate for flavors and fragrances

Formel: C₅H₈O₃
Molekulargewicht: 116.12 g/mol
Siedepunkt: 143…150 °C
Schmelzpunkt: –58 °C
Dichte: 1.045 g/cm³ (25 °C)
Flammpunkt: 45 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00009123
CAS-Nummer: 617-35-6
EINECS: 210-511-2
UN-Nummer: 3272
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR