1-tert-Butoxycarbonyl-4-(cyclopropylamino)piperidine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Cyclopropylamino)piperidine, N1-BOC protected
OR7703-1G
OR7703-250MG
APOSOR7703-1GEA
140
EUR
APOSOR7703-1G
APOSOR7703-250MG
1-tert-Butoxycarbonyl-4-(cyclopropylamino)piperidine 95+%
1-tert-Butoxycarbonyl-4-(cyclopropylamino)piperidine
Formel:
C₁₃H₂₄N₂O₂ Molekulargewicht: 240.35 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03426074 CAS-Nummer: 179557-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...