3'-Formylbiphenyl-2-carbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR7386-1G
OR7386-250MG
APOSOR7386-1GEA
319
EUR
APOSOR7386-1G
APOSOR7386-250MG
3'-Formylbiphenyl-2-carbonsäure
3'-Formylbiphenyl-2-carbonsäure
Formel:
C₁₄H₁₀O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04039067 CAS-Nummer: 100538-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...