4-(Octyloxy)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-n-Octyloxyphenylboronic acid, (4-(Octyloxy)phenyl)boronic acid, 4-n-Octyloxybenzeneboronic acid
OR7245-1G
OR7245-25G
OR7245-5G
APOSOR7245-1GEA
21.9
EUR
APOSOR7245-1G
APOSOR7245-25G
APOSOR7245-5G
4-(Octyloxy)phenylboronsäure
4-(Octyloxy)phenylboronsäure
Formel:
C₁₄H₂₃BO₃ Schmelzpunkt: 88…90 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00982004 CAS-Nummer: 121554-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...