4-(Nonyloxy)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-(Nonyloxy)phenyl)boronic acid

OR7243-1G OR7243-5G
APOSOR7243-1GEA 286 EUR
APOSOR7243-1G APOSOR7243-5G
4-(Nonyloxy)phenylboronsäure 98%
4-(Nonyloxy)phenylboronsäure
Formel: C₁₅H₂₅BO₃
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD04039033
CAS-Nummer: 173392-87-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR