4-(Tetrahydropyran-2-yloxy)phenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR7225-1G
OR7225-5G
APOSOR7225-1GEA
21.9
EUR
APOSOR7225-1G
APOSOR7225-5G
4-(Tetrahydropyran-2-yloxy)phenylboronsäure 97%
4-(Tetrahydropyran-2-yloxy)phenylboronsäure
Formel:
C₁₁H₁₅BO₄ Schmelzpunkt: 116…123 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD02183525 CAS-Nummer: 182281-01-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...