3-Brombenzoylacetonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR7013-5G
OR7013-25G
APOSOR7013-5GEA
161
EUR
APOSOR7013-5G
APOSOR7013-25G
3-Brombenzoylacetonitril
3-Brombenzoylacetonitril
Formel:
C₉H₆BrNO Schmelzpunkt: 93…95 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02260778 CAS-Nummer: 70591-86-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...