1-(2-Cyclohexylethyl)piperazin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6906-25G
OR6906-5G
OR6906-1G
APOSOR6906-25GEA
935
EUR
APOSOR6906-25G
APOSOR6906-5G
APOSOR6906-1G
1-(2-Cyclohexylethyl)piperazin 97%
1-(2-Cyclohexylethyl)piperazin
Formel:
C₁₂H₂₄N₂ Siedepunkt: 98…100 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01320901 CAS-Nummer: 132800-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...