2-(Dimethylamino)benzonitril 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR6854-1G
OR6854-250MG
APOSOR6854-1GEA
102
EUR
APOSOR6854-1G
APOSOR6854-250MG
2-(Dimethylamino)benzonitril 95+%
2-(Dimethylamino)benzonitril
Formel:
C₉H₁₀N₂ Siedepunkt: 130 °C/12mmHg Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD01313389 CAS-Nummer: 20925-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...