5-Brom-2-methoxybenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR6600-1G
OR6600-25G
OR6600-5G
APOSOR6600-1GEA
21.9
EUR
APOSOR6600-1G
APOSOR6600-25G
APOSOR6600-5G
5-Brom-2-methoxybenzonitril
5-Brom-2-methoxybenzonitril
Formel:
C₈H₆BrNO Schmelzpunkt: 91…92 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00143429 CAS-Nummer: 144649-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...